missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Ammonium Cerium(Iv) Nitrate, ACS Reagent, ≥98.5%, Honeywell™ Fluka™

ACS Reagent,≥98.5%

$120.80 - $120.80

Chemical Identifiers

CAS 16774-21-3
MDL Number MFCD00151121
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045239 View Documents Honeywell Fluka™
21547350G
50g Poly Bottle Each for $120.80
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

16774-21-3
MFCD00151121

Specifications

16774-21-3
MFCD00151121
Powder/Crystals
≥98.5%
2.49g/cu.cm
Orange
Acidic
Ammonium Cerium(Iv) Nitrate
Ce(NH4)2(NO3)6
UN3085
548.22g/mol
ACS Reagent
Poly Bottle
214°C
50g
Safety and Handling

Safety and Handling

P210-P220-P260-P280-P305 + P351 + P338-P370 + P378

H272-H290-H302-H314-H317-H410

DOTInformation : Class 5.1 - PG 2 -EHS - acidic - Oxidizing solid, corrosive, n.o.s., HI: all

EINECSNumber : 240-827-6