missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Ammonium Cerium(Iv) Nitrate, ≥98.0% (T), purum p.a., Honeywell™

Purum p.a.,≥98.0% (T)

$903.62 - $903.62

Chemical Identifiers

CAS 16774-21-3
MDL Number MFCD00151121
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045344 View Documents Honeywell™
22250500G
500g Poly Bottle Each for $903.62
 
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications

Chemical Identifiers

16774-21-3
MFCD00151121

Specifications

16774-21-3
MFCD00151121
548.22g/mol
purum p.a.
500g
Ce(NH4)2(NO3)6
UN3085
≥98.0% (T)
Poly Bottle
Ammonium Cerium(Iv) Nitrate
Safety and Handling

Safety and Handling

P210-P220-P260-P280-P305 + P351 + P338-P370 + P378

H272-H290-H302-H314-H317-H410

DOTInformation : Class 5.1 - PG 2 -EHS - acidic - Oxidizing solid, corrosive, n.o.s., HI: all

EINECSNumber : 240-827-6