missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium carbonate, 99.999% (metals basis), Puratronic™, Alfa Aesar™

Manufacturer:  Alfa Aesar 1298009

 View more versions of this product

Catalog No. AA1298009



Specifications

Specifications

Ammonium carbonate
99.999% (metals basis)
MFCD00010890
YNSUIDWLZCFLML-UHFFFAOYSA-N
diazanium;hydrogen carbonate;carbamate
24660
Powder
8000-73-5
C2H11N3O5
ammonium carbonate nf, ammonium carbonate nf, ugam, ammonium bicarbonate-ammonium carbamate, ammonium carbonate chips nf, ammonium bicarbonate, mixt. with ammonium carbamate, ammonium carbamate, mixt. with ammonium bicarbonate, monoammonium carbonate mixture with ammonium carbamate, carbamic acid, monoammonium salt, mixt. with ammonium carbonate, diazanium hydrogen carbonate carbamate
C(=O)(N)[O-].C(=O)(O)[O-].[NH4+].[NH4+]
157.126
96.09
10g

Chemical Identifiers

8000-73-5
157.126
YNSUIDWLZCFLML-UHFFFAOYSA-N
24660
C(=O)(N)[O-].C(=O)(O)[O-].[NH4+].[NH4+]
C2H11N3O5
MFCD00010890
ammonium carbonate nf, ammonium carbonate nf, ugam, ammonium bicarbonate-ammonium carbamate, ammonium carbonate chips nf, ammonium bicarbonate, mixt. with ammonium carbamate, ammonium carbamate, mixt. with ammonium bicarbonate, monoammonium carbonate mixture with ammonium carbamate, carbamic acid, monoammonium salt, mixt. with ammonium carbonate, diazanium hydrogen carbonate carbamate
diazanium;hydrogen carbonate;carbamate
Safety and Handling

Safety and Handling

EINECSNumber : 208-058-0

storageNote1 : Hygroscopic

Documents