Please login to your online account to display your discounted pricing

Ammonium Carbonate (Lumps/NF), Fisher Chemical

Colorless, translucent lumps, 1“ (2.54cm) or smaller in size

$225.62 - $2,776.15

Chemical Identifiers

Chemical Name or Material Ammonium Carbonate
CAS 506-87-6
Purity Grade NF
Physical Form Solid
Assay 30.0 to 34.0%
Assay Percent Range 30.0 to 34.0 %
Molecular Formula CH8N2O3
MDL Number MFCD00010890
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A657-12

  SDS
fisher scientific company
65712
Fiber Drum 12kg Each for $2,776.15

A657-212

  SDS
fisher scientific company
FLA657212
Poly Pail 2.5kg Each for $726.00

A657-500

  SDS
fisher scientific company
FLA657500
Poly Bottle 500g Each for $225.62
Case of 6 Each for $1,353.72
Description & Specifications

Specifications

Chemical Name or Material Ammonium Carbonate
CAS 506-87-6
ChemAlert Storage Symbol Gray
Additional Information Vapor Pressure: 69hPa at 20°C
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:1
Purity Grade NF
Physical Form Solid
Assay 30.0 to 34.0%
Assay Percent Range 30.0 to 34.0 %
Molecular Formula CH8N2O3
MDL Number MFCD00010890
Chloride 0.0035% max.
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Ignition Residue 0.05% max.
Packaging Fiber Drum
Sulfate 0.005% max.
Vapor Pressure 69 hPa at 20°C
Decomposition Information 58°C
Color White
Melting Point 58°C
pH 9.4
Quantity 12kg

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:1

WARNING!