Please login to your online account to display your discounted pricing

Ammonium Carbonate (Lumps/Certified ACS), Fisher Chemical

$236.46 - $236.46

Chemical Identifiers

Chemical Name or Material Ammonium Carbonate
CAS 506-87-6
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥30%
Molecular Formula CH8N2O3
Formula Weight 96.07
MDL Number MFCD00085313
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A656-500

  SDS
fisher scientific company
FLA656500
Poly Bottle 500g Each for $236.46
Case of 6 Each for $1,418.76
Description & Specifications

Specifications

Assay ≥30%
Melting Point 58°C
Purity Grade Certified ACS Grade
Formula Weight 96.07
Quantity 500g
Physical Form Solid
Color White
pH 9.4
Vapor Pressure 69 hPa at 20°C
Decomposition Information 58°C
Merck Index 15, 505
Chloride 5ppm max.
Sulfur Compounds 0.002% max.
Identification Pass Test
Insoluble Matter 0.005% max.
Packaging Poly Bottle
Health Hazard 3 Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:1
Health Hazard 2 WARNING!
ChemAlert Storage Symbol Gray
Chemical Name or Material Ammonium Carbonate
Molecular Formula CH8N2O3
MDL Number MFCD00085313
Non_volatile Matter 0.005% max.
Additional Information Vapor Pressure: 69hPa at 20°C
Iron (Fe) 3ppm max.
Heavy Metals (as Pb) 5ppm max.
CAS 506-87-6

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.