Please login to your online account to display your discounted pricing

Ammonium Carbonate (Lumps/Certified ACS), Fisher BioReagents


This product is a mixture of Ammonium bicarbonate and Ammonium carbonate

Quantity 3kg
Packaging Poly Pail

Manufacturer: fisher scientific company  BP24143

 View more versions of this product

Catalog No. BP24143

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥30%
Purity Grade Certified ACS Grade
Melting Point 58°C
Quantity 3kg
Physical Form Solid
Molecular Formula CH8N2O3
pH 9.4
Color White
Vapor Pressure 69 hPa at 20°C
Chemical Name or Material Ammonium Carbonate
Sulfur Compounds 0.002% max.
Heavy Metals (as Pb) 5ppm max.
Recommended Storage RT
Insoluble Matter 0.005% max.
Chloride 5ppm max.
Iron (Fe) 3ppm max.
Non_volatile Matter 0.005% max.
CAS 8000-73-5
Identification Pass Test
Packaging Poly Pail
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Ammonium Carbonate
CAS 8000-73-5
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥30%
Accessories
 

RT

SDS & Safety Documents