missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Carbonate (Lumps/Certified ACS), Fisher BioReagents

This product is a mixture of Ammonium bicarbonate and Ammonium carbonate

$59.88 - $231.35

Chemical Identifiers

CAS 8000-73-5
Physical Form Solid
Chemical Name or Material Ammonium Carbonate
Assay ≥30%
Grade Certified ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BP24143 SDS Fisher Scientific Company
BP24143
Poly Pail 3kg Each for $231.35
BP2414500 SDS Fisher Scientific Company
BP2414500
Amber Glass 500g Each for $59.88
Case of 6 Each for $359.28
Specifications

Description

Ammonium Carbonate is used in many biochemical procedures.
Description & Specifications

Chemical Identifiers

8000-73-5
Ammonium Carbonate
Certified ACS Grade
Solid
≥30%

Specifications

58°C
Solid
9.4
69 hPa at 20°C
5ppm max.
5ppm max.
0.005% max.
Pass Test
Ammonium Carbonate
Certified ACS Grade
3kg
CH8N2O3
White
0.002% max.
0.005% max.
3ppm max.
8000-73-5
Poly Pail
≥30%

Safety and Handling

Recommended Storage : RT