missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium carbonate, for analysis, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 196650010

 View more versions of this product

Catalog No. AC196650010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ammonium carbonate
506-87-6
(NH4)2CO3
15, 505
Solubility in water: soluble (320g/L at 20°C)
C(=O)([O-])[O-].[NH4+].[NH4+]
96.09
96.09
≥30.0% (NH3)
20ppm max.
10ppm max.
Plastic bottle
50ppm max.
58.0°C
p.a.
CH8N2O3
MFCD00010890
ammonia sesquicarbonate
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
diazanium;carbonate
517111
Crystalline Powder
Analysis
10ppm max.
0.01 % max.
Solubility in water: soluble (320g/L at 20°C).
White
1kg

Chemical Identifiers

506-87-6
96.09
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
517111
C(=O)([O-])[O-].[NH4+].[NH4+]
CH8N2O3
MFCD00010890
ammonia sesquicarbonate
diazanium;carbonate
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-058-

SDS Documents