Please login to your online account to display your discounted pricing

Ammonium Carbonate ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Ammonium Carbonate
CAS 10361-29-2
Purity Grade ACS Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK3352500

 
avantor performance
335204
Glass 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Quantity 500g
CAS 10361-29-2
Packaging Glass
Chemical Name or Material Ammonium Carbonate