missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Ammonium Carbonate, 99.99% Trace Metals Basis, Honeywell™

99.999% trace metals basis

$154.95 - $592.76

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046675 View Documents Honeywell™
37999910G
10g Poly Bottle Each for $154.95
 
60046676 View Documents Honeywell™
37999950G
50g Poly Bottle Each for $592.76
 
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

506-87-6
96.086
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
517111
C(=O)([O-])[O-].[NH4+].[NH4+]
CH8N2O3
MFCD00010890
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
diazanium;carbonate

Specifications

506-87-6
(NH4)2CO3
4253487
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
C(=O)([O-])[O-].[NH4+].[NH4+]
96.086
96.09g/mol
0.9g/cm3 at 20°C
Poly Bottle
69 hPa at 20°C
Temperature : >160°C
9 (at 100g/L 20°C)
Ammonium carbonate
CH8N2O3
MFCD00010890
Solid
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
diazanium;carbonate
517111
99.999% trace metals basis
Ammonia stinging
In water 320g/L at 20°C
2.7 (vs air)
White
10g
Safety and Handling

Safety and Handling

P301 + P312 + P330

RTECSNumber : 208-058-0