missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium carbamate, 98%, Thermo Scientific Chemicals

Ammoniating agent

Supplier:  Thermo Scientific Chemicals 01813430

 View more versions of this product

Catalog No. AA1813430



Description

Description

Ammonium carbamate is used in the preparation of fertilizers. It acts as an ammoniating agent. It plays an important role in the formation of carbamoyl phosphate, which is essential for urea cycle and pyrimidines production. In addition to this, it is used in the preparation of trimethylsilyl carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium carbamate
1111-78-0
250 g
MFCD00013010
ammonium carbamate, ammonium aminoformate, carbamic acid, monoammonium salt, carbamic acid ammonium salt, carbamic acid ammoniate, unii-i2w9615swp, carbamic acid, ammonium salt 1:1, ammoniumcarbamate, anhydride of ammonium carbonate, azanium carbamate
BVCZEBOGSOYJJT-UHFFFAOYSA-N
azanium;carbamate
517232
1.6
60° (sublimation)
Powder
CH6N2O2
3914091
Soluble in cold water and ammonium hydroxide. Slightly soluble in alcohol. Insoluble in acetone.
C(=O)(N)[O-].[NH4+]
78.071
78.07

Chemical Identifiers

1111-78-0
78.071
BVCZEBOGSOYJJT-UHFFFAOYSA-N
517232
C(=O)(N)[O-].[NH4+]
CH6N2O2
MFCD00013010
ammonium carbamate, ammonium aminoformate, carbamic acid, monoammonium salt, carbamic acid ammonium salt, carbamic acid ammoniate, unii-i2w9615swp, carbamic acid, ammonium salt 1:1, ammoniumcarbamate, anhydride of ammonium carbonate, azanium carbamate
azanium;carbamate
Safety and Handling

Safety and Handling

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

EINECSNumber : 214-185-2

RTECSNumber : EY8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only