Please login to your online account to display your discounted pricing

Ammonium Buffer, LabChem™

$102.29 - $102.29

Chemical Identifiers

Chemical Name or Material Ammonium Buffer
CAS 7732-18-5
CAS Max % 89.1
CAS 12125-02-9
CAS Max % 6.6
CAS 1336-21-6
CAS Max % 4.3
Linear Formula NH4Cl
Formula Weight 53.49
UN Number UN2672
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC109712

 
lab chem
LC109712
1L Each for $102.29
Description & Specifications

Specifications

Formula Weight 53.49
Chemical Name or Material Ammonium Buffer
Density 0.94g/mL
Specific Gravity 0.94g/mL
Decomposition Information Gaseous Ammonia; Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
CAS Max % 4.3
Color Clear, colorless
Quantity 1L
Physical Form Liquid
Linear Formula NH4Cl
CAS 7732-18-5
UN Number UN2672
Health Hazard 1 Danger
Identification Passes Test
CAS Max % 89.1
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.
Viscosity 1.27cSt
CAS 12125-02-9
Health Hazard 3 GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 6.6
Recommended Storage Room Temperature
Solubility Soluble in water
CAS 1336-21-6
Safety and Handling

Danger

GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.

Room Temperature