Learn More
Alfa Aesar™ Ammonium bromide, Puratronic™, 99.999% (metals basis)
Manufacturer: Alfa Aesar™ 1298106
Specifications
Ammonium bromide | |
(metals basis) | |
14,505 | |
Crystalline | |
Puratronic™ | |
Hygroscopic | |
452°C (sublimation) |
12124-97-9 | |
MFCD00011419 | |
97.97 | |
99.999% | |
2.429g/mL | |
White | |
5g |
Chemical Identifiers
12124-97-9 | |
MFCD00011419 |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
RTECSNumber : BO9155000
TSCA : TSCA