Please login to your online account to display your discounted pricing

Ammonium Bromide, (Crystalline/ACS Reagent Grade), MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  0219140580

 View more versions of this product

Catalog No. ICN19140580

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Chemical Name or Material Ammonium Bromide
CAS 12124-97-9
Purity Grade ACS Reagent
Physical Form Crystalline
Molecular Formula NH4Br
Formula Weight 97.9g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Ammonium Bromide
CAS 12124-97-9
Purity Grade ACS Reagent
Physical Form Crystalline
Molecular Formula NH4Br
Formula Weight 97.9g/mol
Accessories

Room Temperature