Please login to your online account to display your discounted pricing

Ammonium Bromide, (Crystalline/ACS Reagent Grade), MP Biomedicals

$50.62 - $177.31

Chemical Identifiers

Chemical Name or Material Ammonium Bromide
CAS 12124-97-9
Purity Grade ACS Reagent
Physical Form Crystalline
Molecular Formula NH4Br
Formula Weight 97.9g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19140580

 
mp biomedicals
0219140580
100g Each for $50.62

ICN19140591

 
mp biomedicals
0219140591
1kg Each for $177.31
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Ammonium Bromide
CAS 12124-97-9
Purity Grade ACS Reagent
Physical Form Crystalline
Molecular Formula NH4Br
Formula Weight 97.9g/mol
Recommended Storage Room Temperature
Safety and Handling

Room Temperature