missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Ammonium Bromide, ACS Reagent, ≥99.0%, Honeywell™ Fluka™

ACS Reagent,≥99.0%

$236.06 - $766.53

Chemical Identifiers

CAS 12124-97-9
MDL Number MFCD00011419
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045223 View Documents Honeywell Fluka™
21334925KG
2.5kg Poly Bottle Each for $766.53
 
60045224 View Documents Honeywell Fluka™
213349500G
500g Poly Bottle Each for $236.06
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

12124-97-9
MFCD00011419

Specifications

12124-97-9
MFCD00011419
Crystals
≥99.0%
2.43g/mL at 25°C
≤0.01%
Colorless
5 to 6.5 (at 50g/L 25°C)
Ammonium Bromide
NH4Br
NONH for all modes of transport
97.94g/mol
ACS Reagent
Poly Bottle
1mmHg (198.3°C)
452°C
2.5kg
Safety and Handling

Safety and Handling

P305 + P351 + P338

H319

EINECSNumber : 235-183-8

RTECSNumber : BO9155000