missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium bromide, 99+%, for analysis, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 199980250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ammonium bromide | |
25g | |
BrH4N | |
MFCD00011419 | |
Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C),freely soluble in alcohol and acetone,slightly soluble in ether,practically insoluble in ethyl acetate | |
[NH4+].[Br-] | |
97.94 | |
99+% | |
0.2% (105°C) max. |
12124-97-9 | |
99% min. (on dry substance) (Argentometry) | |
NH4Br | |
15,502 | |
SWLVFNYSXGMGBS-UHFFFAOYSA-N | |
ammonium bromide | |
97.94 | |
Analysis | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 235-183-8
RUO â Research Use Only