Please login to your online account to display your discounted pricing

Ammonium bromide, 99+%, for analysis, ACROS Organics™


Quantity 5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC199980051

 View more versions of this product

Catalog No. AC199980051

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥99.0% (on dry substance) (Argentometry)
Formula Weight 97.94
pH 4.5 to 6.0 (5% soln.)
Quantity 5kg
Color White
Purity Grade For analysis
Chemical Name or Material Ammonium bromide
Heavy Metals (as Pb) 10ppm max.
Nickel (Ni) 2ppm max.
Assay Percent Range 99+%
Sulfated Ash 0.05% max.
CAS 12124-97-9
Copper (Cu) 2ppm max.
Iron (Fe) 1ppm max.
Lead (Pb) 2ppm max.
Loss on Drying 0.2% (105°C) max.
Packaging Plastic bottle
Linear Formula NH4Br
Sulfate 0.005% max.
Health Hazard 1 Warning
Chloride 0.2% max.
Molecular Formula H4BrN
Barium (Ba) 20ppm max.
Bromate 0.001% max.

Specifications

Chemical Name or Material Ammonium bromide
CAS 12124-97-9
Purity Grade For analysis
Assay ≥99.0% (on dry substance) (Argentometry)
Assay Percent Range 99+%
Molecular Formula H4BrN
Linear Formula NH4Br
Formula Weight 97.94
Accessories

Warning

SDS & Safety Documents