missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium bis(oxalato)oxotitanate(IV) hydrate, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1089518

 View more versions of this product

Catalog No. AA1089518



Specifications

Specifications

Ammonium bis(oxalato)oxotitanate(IV) hydrate
(metals basis)
UN3288
276.01 (Anhydrous)
99.998%
Odorless
10580-03-7
MFCD00149620
Ammonium titanyloxalate
Crystalline Powder
Puratronic™
50g

Chemical Identifiers

10580-03-7
MFCD00149620
Ammonium titanyloxalate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents