missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium bicarbonate, 98%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 370930010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium bicarbonate | |
1066-33-7 | |
Crystalline Powder, Crystals and/or Chunks | |
98% | |
NH4HCO3 | |
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 | |
QOPSSYZLVIENBR-UHFFFAOYSA-J | |
dizinc(2+) diammonium tricarbonate | |
14013 | |
98% | |
Authentic |
105°C | |
White | |
1 kg | |
C3H8N2O9Zn2 | |
15,494 | |
Solubility in water: 220g/L (20°C) | |
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O | |
346.86 | |
79.06 | |
Pure | |
Glass bottle |
Chemical Identifiers
1066-33-7 | |
346.86 | |
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 | |
dizinc(2+) diammonium tricarbonate |
C3H8N2O9Zn2 | |
QOPSSYZLVIENBR-UHFFFAOYSA-J | |
14013 | |
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
GHS Signal Word: Warning
EINECSNumber : 213-911-5
RUO – Research Use Only