missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium benzoate, 99%

Manufacturer:  Alfa Aesar™ 3075322

 View more versions of this product

Catalog No. AA3075322



Specifications

Specifications

Ammonium benzoate
99%
MFCD00013069
Benzoic acid ammonium salt
C1=CC=C(C=C1)C(=O)[O-].[NH4+]
139.154
139.15
Weak
White
100g
1863-63-4
C7H9NO2
14,496
VWSRWGFGAAKTQG-UHFFFAOYSA-N
azanium;benzoate
15830
Crystalline
Hygroscopic
192°C to 198°C (decomposition)
110° (230°F)

Chemical Identifiers

1863-63-4
139.154
VWSRWGFGAAKTQG-UHFFFAOYSA-N
15830
C1=CC=C(C=C1)C(=O)[O-].[NH4+]
C7H9NO2
MFCD00013069
Benzoic acid ammonium salt
azanium;benzoate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG3378000

TSCA : TSCA

SDS Documents