Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium aquopentachlororhodate(III), Rh 30% min

Chemical Identifiers

Chemical Name or Material Ammonium aquopentachlororhodate(III)
CAS 63771-33-5
Formula Weight 334.26
MDL Number MFCD00064673
Synonym Ammonium pentachlororhodate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1264503

  SDS
alfa aesar™
1264503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1264506

  SDS
alfa aesar™
1264506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 334.26
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ammonium aquopentachlororhodate(III)
TSCA TSCA
Physical Form Powder
MDL Number MFCD00064673
CAS 63771-33-5
Synonym Ammonium pentachlororhodate
Melting Point 210° to 230°C (dec.)
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Concentration Rh 30% min.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.