missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Acetate (White Crystals), Fisher BioReagents

$133.45 - $187.90

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym acetic acid ammonium salt, acetic acid, ammonium salt, aconh4, ammonium acetate, ammonium ethanoate, ammoniumacetate, azanium acetate, ch3co2nh4, ch3coonh4, unii-rre756s6q2
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP326-500
SDS View Documents Promotion Details
Fisher BioReagents
FLBP326500
500g Poly Bottle
Each for $133.45
Case of 6 Each for $800.70
 
BP326-1
SDS View Documents Promotion Details
Fisher BioReagents
BP3261
1kg Glass Bottle
Each for $187.90
Case of 4 Each for $751.60
 
Description

Description

Ammonium Acetate can be used to precipitate DNA from enzymatic reactions.
Specifications

Chemical Identifiers

631-61-8
77.083
acetic acid ammonium salt, acetic acid, ammonium salt, aconh4, ammonium acetate, ammonium ethanoate, ammoniumacetate, azanium acetate, ch3co2nh4, ch3coonh4, unii-rre756s6q2
CHEBI:62947
CC(=O)[O-].[NH4+]
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate

Specifications

0.01 max. (1M solution) at 280nm
White
7.0
≥97 %
CH3COONH4
15, 492
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
77.08
≥97%
Pass Test
0.005% max.
0.001% max.
0.001% max.
Ammonium Acetate
631-61-8
114°C
500g
C2H7NO2
Pass Test
acetic acid ammonium salt, acetic acid, ammonium salt, aconh4, ammonium acetate, ammonium ethanoate, ammoniumacetate, azanium acetate, ch3co2nh4, ch3coonh4, unii-rre756s6q2
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
Solid
5ppm max.
0.01% max.
5ppm max.
Poly Bottle
5ppm max.
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:1

Recommended Storage : 4°C

SDS
Product Certifications