missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Acetate, ACS Grade, ≥ 97%, LabChem™

$122.74 - $122.74

Chemical Identifiers

CAS 631-61-8
CAS Max % 100
Linear Formula CH3COONH4
Formula Weight 77.08
Chemical Name or Material Ammonium Acetate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC109601 Lab Chem
LC109601
500g Each for $122.74
Specifications Description & Specifications

Chemical Identifiers

631-61-8
CH3COONH4
Ammonium Acetate
100
77.08
ACS Grade

Specifications

77.08
500g
CH3COONH4
Poly Bottle
Crystals
136°C
Room Temperature
≥97%
Gaseous Ammonia; Carbon dioxide; Carbon monoxide; Nitrogen oxides
100
White
Soluble in water
Passes Test
631-61-8
Ammonium Acetate
ACS Grade

Safety and Handling

GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.

GHS P Statement
Avoid breathing dust, vapors.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Keep container tightly closed.
Store locked up and in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

Warning