Please login to your online account to display your discounted pricing

Ammonium Acetate ACS AR,Crystal, Macron Fine Chemicals™


macron

Quantity 125g
Packaging POLYSTORMOR

Manufacturer: avantor performance  327202

 View more versions of this product

Catalog No. MK3272125

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 77.08
Purity Grade ACS Grade
Quantity 125g
Physical Form Crystalline
Chemical Name or Material Ammonium Acetate
CAS 631-61-8
Molecular Formula CH3COONH4
Packaging POLYSTORMOR

Specifications

Chemical Name or Material Ammonium Acetate
CAS 631-61-8
Purity Grade ACS Grade
Physical Form Crystalline
Molecular Formula CH3COONH4
Formula Weight 77.08