Learn More
Ammonium acetate, ACS, 97.0% min, Thermo Scientific Chemicals
Reagent in analytical chemistry, drugs
Supplier: Thermo Scientific Chemicals 011599A3
Description
Ammonium acetate is a reagent used in chromatographic analysis of various compounds such as oligos, proteins, and peptides. It is also employed with acetic acid to provide a buffer solution. Generally it behaves as a catalyst in many reactions like Knoevenagel, Borch. It is also employed as a nutrient additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium acetate | |
631-61-8 | |
2 kg | |
C2H7NO2 | |
4186741 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
azanium;acetate | |
517165 | |
77.08 | |
Hygroscopic |
∽114°C (decomposition) | |
Crystalline | |
97.0% min | |
MFCD00013066 | |
136°C (277°F) | |
Soluble in cold water,methanol ,SO2and acetone. | |
CC(=O)[O-].[NH4+] | |
77.083 | |
CHEBI:62947 | |
ACS | |
1.17 |
Chemical Identifiers
631-61-8 | |
77.083 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
517165 | |
azanium;acetate |
C2H7NO2 | |
MFCD00013066 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CHEBI:62947 | |
CC(=O)[O-].[NH4+] |
Safety and Handling
EINECSNumber : 211-162-9
RTECSNumber : AF3675000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only