missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Acetate - Acetic Acid Solution, TAPPI for Iron, Ricca Chemical

$104.97 - $104.97

Specifications

Chemical Name or Material Ammonium Acetate - Acetic Acid Solution
Color Colorless
CAS Min % 75.0,15.71,7.49
CAS Max % 78.0,16.35,7.79
CAS 64-19-7,631-61-8
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60-216
View Documents Promotion Details
Ricca Chemical Company
RI60216
500 mL Natural Poly Bottle
Each for $104.97
 
Description

Description

Specifications

Specifications

Ammonium Acetate - Acetic Acid Solution
75.0,15.71,7.49
64-19-7,631-61-8
dihydrogen oxide, dihydrogen monoxide
Colorless
78.0,16.35,7.79
Liquid
Natural Poly Bottle
SDS
missing translation for 'documents'

missing translation for 'documents'