Please login to your online account to display your discounted pricing

Ammonium Acetate, 20% (w/v), LabChem™

$94.74 - $94.74

Chemical Identifiers

Chemical Name or Material Ammonium Acetate
CAS 7732-18-5
CAS Max % 80
CAS 631-61-8
CAS Max % 20
Linear Formula CH3COONH4
Formula Weight 77.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC109652

 
lab chem
LC109652
1L Each for $94.74
Description & Specifications

Specifications

Formula Weight 77.08
Chemical Name or Material Ammonium Acetate
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Assay 20% (w/v)
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula CH3COONH4
CAS 7732-18-5
Decomposition Information Gaseous Ammonia; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 1L
Health Hazard 1 Warning
CAS Max % 80
Health Hazard 2 GHS H Statement
Causes skin and serious eye irritation.
May cause respiratory irritation.
CAS 631-61-8
Health Hazard 3 GHS P Statement
Avoid breathing mist.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor/if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Keep container tightly closed.
Store locked up in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 20
Safety and Handling

Warning

GHS P Statement
Avoid breathing mist.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor/if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Keep container tightly closed.
Store locked up in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin and serious eye irritation.
May cause respiratory irritation.