Please login to your online account to display your discounted pricing

Ammonia, T.S., BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$51.56 - $51.56

Chemical Identifiers

Chemical Name or Material Ammonia
Name Note T.S.
Purity Grade BAKER ANALYZED™ Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002262

 
avantor performance
590502
Narrow Mouth Polyethylene Bottle 1L Each for $51.56
Description & Specifications

Specifications

Chemical Name or Material Ammonia
Name Note T.S.
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 0.90kg
Packaging Narrow Mouth Polyethylene Bottle
Quantity 1L