Please login to your online account to display your discounted pricing

Ammonia Standard, Certified, 1000ppm ±10ppm (1mL = 1mg NH3), LabChem™

$51.20 - $51.20

Chemical Identifiers

Chemical Name or Material Ammonia Standard
CAS 7732-18-5
CAS Max % 99.69
CAS 12125-02-9
CAS Max % 0.31
Name Note 1000ppm (1mL = 1mg NH3)
Purity Grade Certified Grade
Linear Formula NH4Cl
Formula Weight 53.49
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC109001

 
lab chem
LC109001
Poly Bottle 500mL Each for $51.20
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 53.49
Chemical Name or Material Ammonia Standard
Specific Gravity 1g/mL
Color Clear, colorless
Quantity 500mL
Decomposition Information Gaseous Ammonia; Hydrogen chloride
Name Note 1000ppm (1mL = 1mg NH3)
Physical Form Liquid
Assay Percent Notes as NH3
Concentration 1000ppm ±10ppm (1mL = 1mg NH3)
Linear Formula NH4Cl
CAS 7732-18-5
Identification Passes Test
CAS Max % 99.69
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 12125-02-9
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS Max % 0.31
Recommended Storage Room Temperature
Density 1g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature