missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Ammonia (IIAR) Pipe Markers, Legend: LIQ/Ammonia/LOW
For use in labeling ammonia piping and its contents. Brady Ammonia (IIAR) Pipe Markers help create a safer, more efficient work environment.
$27.69 - $27.69
Specifications
Color | Black, Orange, Red, Yellow |
---|---|
Display Text | IIAR Abbreviation: (BLANK); Physical State: LIQ; Pressure Level: LOW; Legend: Ammonia |
For Use With (Application) | Pipe and valve marking |
Catalog Number | Mfr. No. | Description | Material | Diameter (Metric) | Diameter (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Material | Diameter (Metric) | Diameter (English) | Price | Quantity | ||
19-110-501
|
Brady™
58009 |
Style II | PVF Over-laminated Polyester (B-689) | 50.8 to 203.2 mm | 2 to 8 in. |
Each for $27.69
|
|
||
Description
- Meets IIAR Standard and is recommended for use in labeling ammonia piping and its contents
- Made of B-689 high performance wrap-around PVF over-laminated polyester and B-681/B-883 polyester over-laminate on fiberglass plastic carrier
Specifications
Black, Orange, Red, Yellow | |
IIAR Abbreviation: (BLANK); Physical State: LIQ; Pressure Level: LOW; Legend: Ammonia | |
Pipe and valve marking |