Please login to your online account to display your discounted pricing

Ammonia-D3 26% solution in D2O, deuteration degree min. 99.5%, for NMR, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$287.13 - $574.07

Chemical Identifiers

Chemical Name or Material Ammonia-D3
Name Note 26% solution in D2O
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula ND4OD
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M8150080010

 
emd millipore corporation
8150080010
Glass ampoule 10mL Each for $287.13

M8150080025

 
emd millipore corporation
8150080025
Glass bottle 25mL Each for $574.07
Description & Specifications

Specifications

Quantity 10mL
Color Colorless
Physical Form Liquid
Chemical Name or Material Ammonia-D3
Vapor Pressure 535 hPa (20°C)
Odor Stinging
Name Note 26% solution in D2O
Assay Percent Range 99.5% min.
Molecular Formula ND4OD
Packaging Glass ampoule