Please login to your online account to display your discounted pricing

Ammonia-D3 26% solution in D2O, deuteration degree min. 99.5%, for NMR, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 25mL
Packaging Glass bottle

Manufacturer: emd millipore corporation  8150080025

 View more versions of this product

Catalog No. M8150080025

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 25mL
Color Colorless
Physical Form Liquid
Chemical Name or Material Ammonia-D3
Vapor Pressure 535 hPa (20°C)
Odor Stinging
Packaging Glass bottle
Name Note 26% solution in D2O
Assay Percent Range 99.5% min.
Molecular Formula ND4OD

Specifications

Chemical Name or Material Ammonia-D3
Name Note 26% solution in D2O
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula ND4OD