missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonia, 2M solution in methanol, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 368398000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ammonia | |
H3N | |
MFCD00011418 | |
05,15 | |
15, 488 | |
800mL | |
N | |
17.03 | |
222 | |
17.03 | |
0.787 |
7664-41-7,67-56-1 | |
NH3 | |
1.90 to 2.20M | |
0.7870g/mL | |
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol | |
QGZKDVFQNNGYKY-UHFFFAOYSA-N | |
azane | |
6°C | |
CHEBI:16134 | |
AcroSeal™ Glass Bottle |
RUO â Research Use Only