missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminopyrazine, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC104490500

 View more versions of this product

Catalog No. AC104490500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder
≥99.0%
Slightly yellow to beige
24, 82
MFCD00006137
Authentic
Aminopyrazine
95.1
50g
117° to 120°C
5049-61-6
Glass bottle
C4H5N3
99+%

Chemical Identifiers

5049-61-6
C4H5N3
MFCD00006137
≥99.0%
99+%
95.1
Aminopyrazine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning