Please login to your online account to display your discounted pricing

Aminopurvalanol A, Tocris

Cyclin-dependent kinase inhibitor

$259.29 - $1,075.94

Chemical Identifiers

Chemical Name or Material Aminopurvalanol A
Assay Percent Range >98%
Molecular Formula C19H26ClN7O
Formula Weight 403.91
Synonym (2R)-2-[[6-[(3-Amino-5-chlorophenyl)amino]-9-(1-methylethyl)-9H-purin-2-yl]amino]-3-methyl-1-butanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

207250

 
r & d systems
207250
50mg Each for $1,075.94

207210

 
r & d systems
207210
10mg Each for $259.29
Description & Specifications

Specifications

Formula Weight 403.91
Quantity 50mg
Chemical Name or Material Aminopurvalanol A
Recommended Storage Store at +4°C
Synonym (2R)-2-[[6-[(3-Amino-5-chlorophenyl)amino]-9-(1-methylethyl)-9H-purin-2-yl]amino]-3-methyl-1-butanol
Molecular Formula C19H26ClN7O
Assay Percent Range >98%
CAS 220792-57-4
Safety and Handling

Store at +4°C