missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminonaphtholsulfonic Acid Solution, Reducing Agent for Silica, Ricca Chemical

$41.67 - $70.27

Chemical Identifiers

CAS 7732-18-5,7631-90-5,7757-83-7,116-63-2
CAS Min % 85.0,12.71,0.42,0.21
CAS Max % 88.0,13.23,0.44,0.22
Physical Form Liquid
Chemical Name or Material Aminonaphtholsulfonic Acid Solution, Reducing Agent for Silica
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
609-24 SDS Ricca Chemical Company
RI609.24
Amber Poly 120mL Each for $41.67
609-216 SDS Ricca Chemical Company
RI609.216
Amber Poly 500mL Each for $70.27
R06092001B Ricca Chemical Company
RIR06092001B
Amber poly 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R06092004B Ricca Chemical Company
RIR06092004B
Black poly 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7631-90-5,7757-83-7,116-63-2
88.0,13.23,0.44,0.22
Aminonaphtholsulfonic Acid Solution, Reducing Agent for Silica
85.0,12.71,0.42,0.21
Liquid

Specifications

Liquid
120mL
88.0,13.23,0.22,0.44
Amber Poly
Clear colorless to slightly colored
85.0,0.42,12.71,0.21
7732-18-5,116-63-2,7631-90-5,7757-83-7
Aminonaphtholsulfonic Acid Solution, Reducing Agent for Silica