missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminonaphtholsulfonic Acid Solution (Amino Solution), ASTM for Phosphorus, Ricca Chemical

$50.91 - $50.91

Chemical Identifiers

CAS 7732-18-5,7681-57-4,7757-83-7,116-63-2
CAS Min % 89.0,5.53,3.3,0.09
CAS Max % 93.0,5.75,3.43,0.09
Physical Form Liquid
Chemical Name or Material Aminonaphtholsulfonic Acid Solution, Amino Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
609-14 Ricca Chemical Company
RI609.14
Amber Poly 120mL Each for $50.91
609-116 Ricca Chemical Company
RI609.116
Amber Poly 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7681-57-4,7757-83-7,116-63-2
93.0,5.75,3.43,0.09
Aminonaphtholsulfonic Acid Solution, Amino Solution
89.0,5.53,3.3,0.09
Liquid

Specifications

7732-18-5,7681-57-4,7757-83-7,116-63-2
93.0,5.75,3.43,0.09
Amber Poly
Clear colorless to slightly colored
Aminonaphtholsulfonic Acid Solution, Amino Solution
89.0,5.53,3.3,0.09
Liquid
ASTM for Phosphorus
120mL