Learn More
(Aminomethyl)cyclopropane, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC258720500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(Aminomethyl)cyclopropane | |
0.8200g/mL | |
84.0°C to 85.0°C | |
96% min. (GC) | |
C4H9N | |
1.4320 to 1.4340 | |
50g | |
0.82 | |
cyclopropylmethanamine | |
75646 | |
Liquid |
2516-47-4 | |
Colorless to Yellow | |
−30°C | |
Authentic | |
Glass bottle | |
1-cyclopropylmethanamine, aminomethyl cyclopropane, aminomethylcyclopropane, cyclopropane methyl amine, cyclopropanemethanamine, cyclopropanemethylamine, cyclopropanmethylamine, cyclopropylmethylamin, cyclopropylmethylamine | |
IGSKHXTUVXSOMB-UHFFFAOYSA-N | |
C1CC1CN | |
71.11 | |
71.11 | |
97% |
Chemical Identifiers
2516-47-4 | |
71.11 | |
1-cyclopropylmethanamine, aminomethyl cyclopropane, aminomethylcyclopropane, cyclopropane methyl amine, cyclopropanemethanamine, cyclopropanemethylamine, cyclopropanmethylamine, cyclopropylmethylamin, cyclopropylmethylamine | |
cyclopropylmethanamine |
C4H9N | |
IGSKHXTUVXSOMB-UHFFFAOYSA-N | |
75646 | |
C1CC1CN |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 219-737-6
RUO – Research Use Only