Please login to your online account to display your discounted pricing

Aminomethyl polymer resin, 1% crossl. with DVB, 0.5-0.7mmol/g, 200-400 mesh, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  349370250

 View more versions of this product

Catalog No. AC349370250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 25g
Color Off-white to light yellow
Additional Information crosslinking: 1% DVB
Chemical Name or Material Aminomethyl polymer resin
MDL Number MFCD01092588
CAS 78578-28-6
Synonym Aminomethylpolystyrene divinylbenzene copolymer
Loading 0.5 to 0.7mmol/g
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Mesh Size 200 to 400 mesh

Specifications

Chemical Name or Material Aminomethyl polymer resin
CAS 78578-28-6
MDL Number MFCD01092588
Synonym Aminomethylpolystyrene divinylbenzene copolymer
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents