missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminomethyl polymer resin, 1% crossl. with DVB, 0.5-0.7mmol/g, 200-400 mesh, Acros Organics

Chemical Identifiers

CAS 78578-28-6
MDL Number MFCD01092588
Synonym Aminomethylpolystyrene divinylbenzene copolymer
Chemical Name or Material Aminomethyl polymer resin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC349370050 SDS Thermo Fisher Scientific
349370050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC349370250 SDS Thermo Fisher Scientific
349370250
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

for solid support synthesis
Description & Specifications

Chemical Identifiers

78578-28-6
MFCD01092588
Aminomethylpolystyrene divinylbenzene copolymer
Aminomethyl polymer resin

Specifications

crosslinking: 1% DVB
0.5 to 0.7mmol/g
5g
Beads
Aminomethylpolystyrene divinylbenzene copolymer
78578-28-6
200 to 400 mesh
Off-white to light yellow
MFCD01092588
Aminomethyl polymer resin

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.