Please login to your online account to display your discounted pricing

Aminomethyl polymer resin, 1% crossl. with DVB, 0.4-0.6mmol/g, 100-200 mesh, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  349340050

 View more versions of this product

Catalog No. AC349340050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 5g
Color Off-white to light yellow
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym Aminomethylpolystyrene divinylbenzene copolymer
Chemical Name or Material Aminomethyl polymer resin
Additional Information crosslinking: 1% DVB
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
CAS 78578-28-6
MDL Number MFCD00147824
Loading 0.4 to 0.6mmol/g
Mesh Size 100 to 200 mesh

Specifications

Chemical Name or Material Aminomethyl polymer resin
CAS 78578-28-6
MDL Number MFCD00147824
Synonym Aminomethylpolystyrene divinylbenzene copolymer
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents