Learn More
(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L09833MD
Description
(Aminomethyl)phosphonic acid is used in the preparation of (acetylamino-methyl)-phosphonic acid by reacting with acetic anhydride. Further, it is used in the development of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Aminomethyl)phosphonic acid is used in the preparation of (acetylamino-methyl)-phosphonic acid by reacting with acetic anhydride. Further, it is used in the development of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong bases and oxidizing agents.
Specifications
(Aminomethyl)phosphonic acid | |
∽290°C (decomposition) | |
MFCD00008105 | |
1701219 | |
aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl | |
MGRVRXRGTBOSHW-UHFFFAOYSA-N | |
aminomethylphosphonic acid | |
14017 | |
111.04 |
1066-51-9 | |
CH6NO3P | |
250 mg | |
Hygroscopic | |
Soluble in water. | |
C(N)P(=O)(O)O | |
111.037 | |
CHEBI:28812 | |
99% |
Chemical Identifiers
1066-51-9 | |
111.037 | |
MGRVRXRGTBOSHW-UHFFFAOYSA-N | |
14017 | |
aminomethylphosphonic acid |
CH6NO3P | |
MFCD00008105 | |
aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl | |
CHEBI:28812 | |
C(N)P(=O)(O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 000-000-0
RTECSNumber : SZ6526000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only