missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Aminomethyl)cyclopropane hydrochloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC104010010

 View more versions of this product

Catalog No. AC104010010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(Aminomethyl)cyclopropane hydrochloride
7252-53-1
96.0
White
1g
C4H9N·HCl
107.58
42.87 to 46.44% C, 12.50 to 13.54% N
Authentic
100.0
Powder
97%
C3H5CH2NH2·HCl
Cyclopropanemethylamine hydrochloride

Chemical Identifiers

7252-53-1
100.0
C4H9N·HCl
107.58
(Aminomethyl)cyclopropane hydrochloride
96.0
97%
C3H5CH2NH2·HCl
Cyclopropanemethylamine hydrochloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.