missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Aminomethyl)cyclopropane hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 7252-53-1
Molecular Formula C4H10ClN
Molecular Weight (g/mol) 107.581
InChI Key HXSNGAHNYZBZTH-UHFFFAOYSA-N
Synonym 1-cyclopropylmethanamine hydrochloride
PubChem CID 81671
IUPAC Name cyclopropylmethanamine;hydrochloride
SMILES C1CC1CN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC104010010 SDS View Documents Acros Organics
AC104010010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC104010100 SDS View Documents Acros Organics
AC104010100
10g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7252-53-1
107.581
1-cyclopropylmethanamine hydrochloride
cyclopropylmethanamine;hydrochloride
C4H10ClN
HXSNGAHNYZBZTH-UHFFFAOYSA-N
81671
C1CC1CN.Cl

Specifications

42.87 to 46.44% C, 12.50 to 13.54% N
White
1g
96.0
97%
C3H5CH2NH2·HCl
HXSNGAHNYZBZTH-UHFFFAOYSA-N
cyclopropylmethanamine;hydrochloride
81671
Powder
(Aminomethyl)cyclopropane hydrochloride
Authentic
205.0°C to 209.0°C
7252-53-1
100.0
C4H10ClN
1-cyclopropylmethanamine hydrochloride
C1CC1CN.Cl
107.581
107.58
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.