missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Aminomethyl)cyclopropane hydrochloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC104010100

 View more versions of this product

Catalog No. AC104010100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(Aminomethyl)cyclopropanehydrochloride
Authentic
205.0°C to 209.0°C
7252-53-1
100.0
C4H9N·HCl
12,4
HXSNGAHNYZBZTH-UHFFFAOYSA-N
cyclopropylmethanamine;hydrochloride
81671
Powder
42.87 to 46.44% C, 12.50 to 13.54% N
White
10g
96.0
97%
C3H5CH2NH2·HCl
1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, aminomethylcyclopropane hydrochloride, cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethanamine-hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethylamine hydrochloride
C1CC1CN.Cl
107.58
107.58
97%

Chemical Identifiers

7252-53-1
107.58
1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, aminomethylcyclopropane hydrochloride, cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethanamine-hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethylamine hydrochloride
cyclopropylmethanamine;hydrochloride
C4H9N·HCl
HXSNGAHNYZBZTH-UHFFFAOYSA-N
81671
C1CC1CN.Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-668-

SDS Documents