missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminomethanesulfonic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC155745000

 View more versions of this product

Catalog No. AC155745000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Aminomethanesulfonic acid
Glass bottle
180.0°C to 185.0°C
97%
97%
H2NCH2SO3H
01,583
OBESRABRARNZJB-UHFFFAOYSA-N
aminomethanesulfonic acid
83791
Fine Powder
Authentic
White
500g
13881-91-9
CH5NO3S
MFCD00008124
aminomethanesulfonic, aminomethanesulfonicacid, aminomethanesulfoniccid, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, methanesulfonic acid, 1-amino, methanesulfonic acid, amino, sodium aminomethanesulphonate, unii-ola224z482
C(N)S(=O)(=O)O
111.12
111.12
97%

Chemical Identifiers

13881-91-9
111.12
OBESRABRARNZJB-UHFFFAOYSA-N
83791
C(N)S(=O)(=O)O
CH5NO3S
MFCD00008124
aminomethanesulfonic, aminomethanesulfonicacid, aminomethanesulfoniccid, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, methanesulfonic acid, 1-amino, methanesulfonic acid, amino, sodium aminomethanesulphonate, unii-ola224z482
aminomethanesulfonic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 237-649-6

TSCA : TSCA

SDS Documents