Please login to your online account to display your discounted pricing

Alfa Aesar™ Aminoguanidine sulfate, 98%

$226.01 - $226.01

Chemical Identifiers

Chemical Name or Material Aminoguanidine sulfate
CAS 1068-42-4
Molecular Formula C2H12N8·H2SO4
Formula Weight 246.24
MDL Number MFCD00151369
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA197100B

  SDS
alfa aesar™
A197100B
1000g Each for $226.01
Description & Specifications

Specifications

Formula Weight 246.24
Quantity 1000g
MDL Number MFCD00151369
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C2H12N8·H2SO4
Assay 98%
CAS 1068-42-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material Aminoguanidine sulfate
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.