missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminoguanidine nitrate, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC103760050

 View more versions of this product

Catalog No. AC103760050



Specifications

Specifications

Aminoguanidine nitrate
Glass bottle
145°C to 147°C
99%
99%
H2NNHC(=NH)NH2·HNO3
03,117
PMGFHEJUUBDCLU-UHFFFAOYSA-N
2-aminoguanidine;nitric acid
165859
Fine Crystalline Powder
Authentic
White
5g
10308-82-4
CH6N4·HNO3
MFCD00013174
1-aminoguanidine; nitric acid
C(=NN)(N)N.[N+](=O)(O)[O-]
137.11
137.11
≥98.5% (Total base)

Chemical Identifiers

10308-82-4
137.11
PMGFHEJUUBDCLU-UHFFFAOYSA-N
165859
C(=NN)(N)N.[N+](=O)(O)[O-]
CH6N4·HNO3
MFCD00013174
1-aminoguanidine; nitric acid
2-aminoguanidine;nitric acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-682-5

TSCA : TSCA

SDS Documents