missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminoguanidine nitrate, 98%, cont. ca 10% water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1163214

 View more versions of this product

Catalog No. AAL1163214



Description

Description

Aminoguanidine nitrate is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine nitrate is used as a pharmaceutical raw material.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with reducing agents, strong oxidizing agents, organic materials and metal powders.
Specifications

Specifications

Aminoguanidine nitrate
138°C to 142°C
CH7N5O3
25 g
3916056
Soluble in water.
O[N+]([O-])=O.NN=C(N)N
137.10
137.1
10308-82-4
cont. ≈10% water
MFCD00013174
UN1479
aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
PMGFHEJUUBDCLU-UHFFFAOYSA-N
2-aminoguanidine;nitric acid
165859
98%

Chemical Identifiers

10308-82-4
137.10
PMGFHEJUUBDCLU-UHFFFAOYSA-N
165859
O[N+]([O-])=O.NN=C(N)N
CH7N5O3
MFCD00013174
aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
2-aminoguanidine;nitric acid
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 233-682-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only